Assay Details
Target Gene Details
Entrez Gene ID: | 221188 |
Gene Name: | adhesion G protein-coupled receptor G5 |
Gene Aliases: |
GPR114, PGR27 |
Location: |
Chr.16:57529339-57578182 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRG5 | NM_001304376.2 | NP_001291305.1 | ||
| NM_001318481.1 | NP_001305410.1 | |||
| NM_153837.3 | NP_722579.1 | |||
| XM_011522949.2 | XP_011521251.1 | |||
| XM_011522950.2 | XP_011521252.1 | |||
| AK128298.1 | ||||
| AK160368.1 | BAD18711.1 | |||
| AK292783.1 | ||||
| AK301357.1 | ||||
| AK303661.1 | ||||
| AY358447.1 | AAQ88812.1 | |||
| BC032401.1 | AAH32401.1 | |||
| BM193863.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833245 | Chr.16:57434073 - 57584619 on Build GRCh38 | Loss |
|
| nsv1055537 | Chr.16:57531862 - 57570612 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map