Assay Details
Target Gene Details
Entrez Gene ID: | 23090 |
Gene Name: | zinc finger protein 423 |
Gene Aliases: |
Ebfaz, JBTS19, NPHP14, OAZ, Roaz, ZFP423, Zfp104, hOAZ |
Location: |
Chr.16:49487531-49857919 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF423 | NM_001271620.2 | NP_001258549.1 | ||
| NM_015069.4 | NP_055884.2 | |||
| XM_005255856.4 | XP_005255913.1 | |||
| XM_005255857.4 | XP_005255914.1 | |||
| XM_006721171.3 | XP_006721234.1 | |||
| XM_017023076.1 | XP_016878565.1 | |||
| XM_017023077.1 | XP_016878566.1 | |||
| XM_017023078.1 | XP_016878567.1 | |||
| AB018303.2 | BAA34480.2 | |||
| AF221712.1 | AAF28354.1 | |||
| AK027479.1 | ||||
| AK304829.1 | ||||
| AK309835.1 | ||||
| AK314742.1 | ||||
| BC112315.1 | AAI12316.1 | |||
| BC112317.1 | AAI12318.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833222 | Chr.16:49367505 - 49536359 on Build GRCh38 | Loss |
|
| nsv522048 | Chr.16:49510329 - 49520969 on Build GRCh38 | Loss |
|
| nsv833223 | Chr.16:49496517 - 49687323 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs147878172] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map