Assay Details
Target Gene Details
Entrez Gene ID: | 8883 |
Gene Name: | NEDD8 activating enzyme E1 subunit 1 |
Gene Aliases: |
A-116A10.1, APPBP1, HPP1, ula-1 |
Location: |
Chr.16:66802878-66830977 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAE1 | NM_001018159.1 | NP_001018169.1 | ||
| NM_001018160.1 | NP_001018170.1 | |||
| NM_001286500.1 | NP_001273429.1 | |||
| NM_003905.3 | NP_003896.1 | |||
| XM_005256215.1 | XP_005256272.1 | |||
| XM_011523422.2 | XP_011521724.1 | |||
| AK097680.1 | ||||
| AK298159.1 | ||||
| AK312784.1 | ||||
| AL136798.1 | CAB66732.1 | |||
| AY197612.1 | AAP35030.1 | |||
| BC000480.2 | AAH00480.1 | |||
| BC013301.2 | AAH13301.1 | |||
| U50939.1 | AAC50477.1 |
More Information
Set Membership: |
|

Genomic Map