Assay Details
Target Gene Details
Entrez Gene ID: | 57478 |
Gene Name: | ubiquitin specific peptidase 31 |
Gene Aliases: |
- |
Location: |
Chr.16:23061407-23149301 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP31 | NM_020718.3 | 16 | 10060 | NP_065769.3 |
| XM_017023496.1 | 15 | 9971 | XP_016878985.1 | |
| XM_017023497.1 | 16 | 11586 | XP_016878986.1 | |
| XM_017023498.1 | 15 | 9373 | XP_016878987.1 | |
| XM_017023499.1 | 13 | 9177 | XP_016878988.1 |
More Information
Set Membership: |
|

Genomic Map