Assay Details
Target Gene Details
Entrez Gene ID: | 65009 |
Gene Name: | NDRG family member 4 |
Gene Aliases: |
BDM1, SMAP-8, SMAP8 |
Location: |
Chr.16:58463645-58515403 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDRG4 | NM_001130487.1 | NP_001123959.1 | ||
| NM_020465.3 | NP_065198.1 | |||
| NM_022910.3 | 1 | 100 | NP_075061.1 | |
| XM_006721252.3 | XP_006721315.1 | |||
| XM_006721254.3 | XP_006721317.1 | |||
| XM_006721256.3 | XP_006721319.1 | |||
| XM_006721257.3 | XP_006721320.1 | |||
| XM_006721258.3 | XP_006721321.1 | |||
| XM_006721260.3 | XP_006721323.1 | |||
| XM_011523290.2 | 1 | 100 | XP_011521592.1 | |
| XM_011523291.2 | 1 | 100 | XP_011521593.1 | |
| XM_011523292.2 | 1 | 100 | XP_011521594.1 | |
| XM_011523293.2 | 1 | 148 | XP_011521595.1 | |
| XM_011523294.2 | XP_011521596.1 | |||
| XM_017023582.1 | 1 | 100 | XP_016879071.1 | |
| XM_017023583.1 | 1 | 100 | XP_016879072.1 | |
| XM_017023584.1 | XP_016879073.1 | |||
| XM_017023585.1 | XP_016879074.1 | |||
| XM_017023587.1 | XP_016879076.1 | |||
| XM_017023588.1 | XP_016879077.1 | |||
| XM_017023590.1 | XP_016879079.1 | |||
| AB021172.1 | BAB20288.1 | |||
| AK126574.1 | BAC86600.1 | |||
| AK126729.1 | ||||
| CF552009.1 | ||||
| CR936733.1 | ||||
| DA069318.1 | ||||
| DA099057.1 | 2 | 255 | ||
| DA242460.1 | ||||
| DB498690.1 |
More Information
Set Membership: |
|

Genomic Map