Assay Details
Target Gene Details
Entrez Gene ID: | 8448 |
Gene Name: | double C2 domain alpha |
Gene Aliases: |
Doc2 |
Location: |
Chr.16:30005514-30023280 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOC2A | NM_001282062.1 | NP_001268991.1 | ||
| NM_001282068.1 | NP_001268997.1 | |||
| XM_011545975.1 | XP_011544277.1 | |||
| XM_017023776.1 | XP_016879265.1 | |||
| XM_017023777.1 | XP_016879266.1 | |||
| BC063436.1 | AAH63436.1 | |||
| BI757951.1 | ||||
| D31897.1 | BAA06695.1 | |||
| DC399462.1 |
More Information
Set Membership: |
|

Genomic Map