Assay Details
Target Gene Details
Entrez Gene ID: | 1012 |
Gene Name: | cadherin 13 |
Gene Aliases: |
CDHH, P105 |
Location: |
Chr.16:82626794-83796610 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH13 | NM_001220488.1 | NP_001207417.1 | ||
| NM_001220489.1 | NP_001207418.1 | |||
| NM_001220490.1 | NP_001207419.1 | |||
| NM_001220491.1 | NP_001207420.1 | |||
| NM_001220492.1 | NP_001207421.1 | |||
| NM_001257.4 | NP_001248.1 | |||
| XM_017022848.1 | XP_016878337.1 | |||
| XM_017022849.1 | XP_016878338.1 | |||
| AK294277.1 | ||||
| AK295886.1 | ||||
| AK298612.1 | ||||
| AK304592.1 | ||||
| AK304733.1 | ||||
| AK307862.1 | ||||
| AU123178.1 | ||||
| BC028624.2 | AAH28624.1 | |||
| BC030653.2 | AAH30653.1 | |||
| BI756586.1 | ||||
| CV024366.1 | ||||
| DC376661.1 | ||||
| EU190357.1 | ||||
| EU190358.1 | ||||
| L34058.1 | AAA35624.1 | |||
| U59288.1 | AAB18911.1 |
Target Gene Details
Entrez Gene ID: | 101928417 |
Gene Name: | uncharacterized LOC101928417 |
Gene Aliases: |
- |
Location: |
Chr.16:82953230-82990298 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928417 | NR_110937.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5255n54 | Chr.16:82955372 - 83011060 on Build GRCh38 | Loss |
|
| nsv573371 | Chr.16:82131578 - 83629456 on Build GRCh38 | Loss |
|
| nsv573367 | Chr.16:82106956 - 83007241 on Build GRCh38 | Gain |
|
| nsv1061288 | Chr.16:82146152 - 83636829 on Build GRCh38 | Gain+Loss |
|
| dgv3051n100 | Chr.16:82138730 - 83654472 on Build GRCh38 | Gain |
|
| esv3582395 | Chr.16:82954267 - 83021814 on Build GRCh38 | Loss |
|
| dgv5253n54 | Chr.16:82148499 - 83630977 on Build GRCh38 | Gain |
|
| esv3892918 | Chr.16:82916897 - 83009568 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75556976] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map