Assay Details
Target Gene Details
Entrez Gene ID: | 3931 |
Gene Name: | lecithin-cholesterol acyltransferase |
Gene Aliases: |
- |
Location: |
Chr.16:67939884-67944112 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCAT | NM_000229.1 | NP_000220.1 | ||
| BC014781.1 | AAH14781.1 | |||
| BT009748.1 | AAP88750.1 | |||
| M12625.1 | AAA59498.1 | |||
| M26268.1 | AAA59499.1 | |||
| X06537.1 | CAB56610.1 |
Target Gene Details
Entrez Gene ID: | 6560 |
Gene Name: | solute carrier family 12 member 4 |
Gene Aliases: |
CTC-479C5.17, KCC1, hKCC1 |
Location: |
Chr.16:67943474-67968694 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC12A4 | NM_001145961.1 | 24 | 4478 | NP_001139433.1 |
| NM_001145962.1 | 23 | 4401 | NP_001139434.1 | |
| NM_001145963.1 | 24 | 4375 | NP_001139435.1 | |
| NM_001145964.1 | 24 | 4366 | NP_001139436.1 | |
| NM_005072.4 | 24 | 4496 | NP_005063.1 | |
| AI368074.1 | 1 | 278 | ||
| AK310027.1 | 1 | 778 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map