Assay Details
Target Gene Details
Entrez Gene ID: | 57478 |
Gene Name: | ubiquitin specific peptidase 31 |
Gene Aliases: |
- |
Location: |
Chr.16:23061407-23149301 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP31 | NM_020718.3 | NP_065769.3 | ||
| XM_017023496.1 | XP_016878985.1 | |||
| XM_017023497.1 | XP_016878986.1 | |||
| XM_017023498.1 | XP_016878987.1 | |||
| XM_017023499.1 | XP_016878988.1 | |||
| XM_017023500.1 | XP_016878989.1 | |||
| AJ586135.2 | CAE51935.2 | |||
| AK126752.1 | ||||
| BX648357.1 | CAH10383.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521703 | Chr.16:23074311 - 23103528 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map