Assay Details
Target Gene Details
Entrez Gene ID: | 65009 |
Gene Name: | NDRG family member 4 |
Gene Aliases: |
BDM1, SMAP-8, SMAP8 |
Location: |
Chr.16:58463645-58515403 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
More Information
Set Membership: |
|

Genomic Map