Assay Details
Target Gene Details
Entrez Gene ID: | 55512 |
Gene Name: | sphingomyelin phosphodiesterase 3 |
Gene Aliases: |
NSMASE2 |
Location: |
Chr.16:68358327-68448703 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMPD3 | NM_018667.3 | NP_061137.1 | ||
| XM_005256031.3 | XP_005256088.1 | |||
| XM_005256032.3 | XP_005256089.1 | |||
| XM_011523207.1 | XP_011521509.1 | |||
| XM_011523208.2 | XP_011521510.1 | |||
| XM_011523209.2 | XP_011521511.1 | |||
| XM_017023405.1 | XP_016878894.1 | |||
| XM_017023406.1 | XP_016878895.1 | |||
| XM_017023407.1 | XP_016878896.1 | |||
| XM_017023408.1 | XP_016878897.1 | |||
| AJ250460.1 | CAB92964.1 | |||
| AK289651.1 | ||||
| BC112238.1 | AAI12239.1 | |||
| BC143631.1 | ||||
| BC143634.1 | ||||
| DQ424899.1 | ABD83664.1 |
More Information
Set Membership: |
|

Genomic Map