Assay Details
Target Gene Details
Entrez Gene ID: | 84662 |
Gene Name: | GLIS family zinc finger 2 |
Gene Aliases: |
NKL, NPHP7 |
Location: |
Chr.16:4314761-4339597 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLIS2 | NM_001318918.1 | NP_001305847.1 | ||
| XM_005255641.4 | XP_005255698.1 | |||
| AK126918.1 |
Target Gene Details
Entrez Gene ID: | 101926896 |
Gene Name: | GLIS2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.16:4324648-4328340 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLIS2-AS1 | NR_110901.1 | |||
| BX104227.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv571300 | Chr.16:4317729 - 4337423 on Build GRCh38 | Loss |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| nsv571282 | Chr.16:4315234 - 4338138 on Build GRCh38 | Gain |
|
| nsv527866 | Chr.16:4206462 - 4412618 on Build GRCh38 | Loss |
|
| nsv571274 | Chr.16:4309420 - 4441189 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map