Assay Details
Target Gene Details
Entrez Gene ID: | 11269 |
Gene Name: | DEAD-box helicase 19B |
Gene Aliases: |
DBP5, DDX19, RNAh |
Location: |
Chr.16:70289767-70335213 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Gene Details
Entrez Gene ID: | 100506083 |
Gene Name: | uncharacterized LOC100506083 |
Gene Aliases: |
- |
Location: |
Chr.16:70315640-70346747 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506083 | NR_039997.1 |
More Information
Set Membership: |
|

Genomic Map