Assay Details
Target Gene Details
Entrez Gene ID: | 28987 |
Gene Name: | NIN1/PSMD8 binding protein 1 homolog |
Gene Aliases: |
ART-4, MST158, MSTP158, NOB1P, PSMD8BP1 |
Location: |
Chr.16:69741854-69754968 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOB1 | NM_014062.2 | NP_054781.1 | ||
| NR_074074.1 | ||||
| AB026125.1 | BAA86961.1 | |||
| AF190161.1 | AAQ13705.1 | |||
| AK001028.1 | BAA91473.1 | |||
| AK314307.1 | ||||
| AY487344.1 | AAR85357.1 | |||
| BC000050.2 | AAH00050.2 | |||
| BC064630.1 | AAH64630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524364 | Chr.16:69711242 - 69769540 on Build GRCh38 | Loss |
|
| esv3638931 | Chr.16:69738369 - 69799862 on Build GRCh38 | Gain |
|
| esv3892878 | Chr.16:69687512 - 69844258 on Build GRCh38 | Gain |
|
| esv3638928 | Chr.16:69677077 - 69756070 on Build GRCh38 | Gain |
|
| esv3638930 | Chr.16:69735814 - 69773243 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map