Assay Details
Target Gene Details
Entrez Gene ID: | 84937 |
Gene Name: | zinc and ring finger 1, E3 ubiquitin protein ligase |
Gene Aliases: |
NIN283 |
Location: |
Chr.16:74999017-75110994 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNRF1 | NM_032268.4 | NP_115644.1 | ||
| XM_011523392.1 | XP_011521694.1 | |||
| XM_017023793.1 | XP_016879282.1 | |||
| AF378524.1 | AAK69753.1 | |||
| AK294434.1 | ||||
| AL136903.1 | CAB66837.1 | |||
| AL834440.1 | CAD39100.1 | |||
| BC007235.1 | AAH07235.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3639081 | Chr.16:75036890 - 75056954 on Build GRCh38 | Gain |
|
| nsv833280 | Chr.16:74821913 - 75043329 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114218249] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map