Assay Details
Target Gene Details
Entrez Gene ID: | 79585 |
Gene Name: | coronin 7 |
Gene Aliases: |
0610011B16Rik, CRN7, POD1 |
Location: |
Chr.16:4354542-4416961 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORO7 | NM_001201472.1 | NP_001188401.1 | ||
| NM_001201473.1 | NP_001188402.1 | |||
| NM_024535.4 | NP_078811.3 | |||
| AK025674.1 | BAB15211.1 | |||
| AK094596.1 | ||||
| AK097238.1 | ||||
| AK294045.1 | ||||
| AK296723.1 | ||||
| AK296807.1 | ||||
| AK303353.1 | ||||
| AK309327.1 | ||||
| AK309763.1 | ||||
| BC117289.1 | ||||
| BC117291.1 |
Target Gene Details
Entrez Gene ID: | 100529144 |
Gene Name: | CORO7-PAM16 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:4340251-4416961 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORO7-PAM16 | NM_001201479.1 | NP_001188408.1 | ||
| AL833954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471075 | Chr.16:4317729 - 4395326 on Build GRCh38 | Loss |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| nsv527866 | Chr.16:4206462 - 4412618 on Build GRCh38 | Loss |
|
| nsv571274 | Chr.16:4309420 - 4441189 on Build GRCh38 | Gain |
|
| esv3584924 | Chr.16:4330628 - 4469258 on Build GRCh38 | Gain |
|
| nsv520212 | Chr.16:4354619 - 4390827 on Build GRCh38 | Loss |
|
| esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs139550185] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map