Assay Details
Target Gene Details
Entrez Gene ID: | 81691 |
Gene Name: | exonuclease NEF-sp |
Gene Aliases: |
- |
Location: |
Chr.16:20806429-20849668 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC81691 | NM_001144924.1 | NP_001138396.1 | ||
| NM_001199053.1 | NP_001185982.1 | |||
| NM_030941.2 | NP_112203.2 | |||
| XM_005255604.1 | XP_005255661.1 | |||
| XM_011545963.1 | XP_011544265.1 | |||
| XM_011545964.1 | XP_011544266.1 | |||
| XM_011545965.1 | XP_011544267.1 | |||
| XM_011545966.1 | XP_011544268.1 | |||
| XM_017023739.1 | XP_016879228.1 | |||
| AF332193.1 | AAK17192.1 | |||
| AK314458.1 | ||||
| AL136763.2 | CAB66697.1 | |||
| BC007646.1 | AAH07646.1 | |||
| BC068503.1 | AAH68503.1 | |||
| DB030041.1 | ||||
| DB059505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050009 | Chr.16:20773077 - 20862801 on Build GRCh38 | Loss |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
| nsv833165 | Chr.16:20718218 - 20886396 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map