Assay Details
Target Gene Details
Entrez Gene ID: | 3294 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 2 |
Gene Aliases: |
EDH17B2, HSD17, SDR9C2 |
Location: |
Chr.16:82035237-82098534 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSD17B2 | NM_002153.2 | NP_002144.1 | ||
| AK223001.1 | BAD96721.1 | |||
| AK313109.1 | ||||
| AU139034.1 | ||||
| BC009581.1 | AAH09581.1 | |||
| BC059170.1 | AAH59170.1 | |||
| BP270993.1 | ||||
| L11708.1 | AAA03562.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs117063842] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map