Assay Details
Target Gene Details
Entrez Gene ID: | 5579 |
Gene Name: | protein kinase C beta |
Gene Aliases: |
PKC-beta, PKCB, PRKCB1, PRKCB2 |
Location: |
Chr.16:23835979-24220611 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCB | NM_002738.6 | NP_002729.2 | ||
| NM_212535.2 | NP_997700.1 | |||
| BC036472.1 | AAH36472.1 | |||
| BI549165.1 | ||||
| DB456157.1 | ||||
| M13975.1 | AAA60095.1 | |||
| X06318.1 | CAA29634.1 | |||
| X07109.1 | CAA30130.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73537024] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map