Assay Details
Target Gene Details
Entrez Gene ID: | 100507458 |
Gene Name: | ZNF213 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.16:3125529-3134882 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF213-AS1 | NR_110900.1 | |||
| AF098968.1 | ||||
| BG752676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509589 | Chr.16:3045935 - 3169200 on Build GRCh38 | Insertion |
|
| nsv571249 | Chr.16:3115424 - 3251897 on Build GRCh38 | Gain |
|
| nsv571248 | Chr.16:3034108 - 3165103 on Build GRCh38 | Loss |
|

Genomic Map