Assay Details
Target Gene Details
Entrez Gene ID: | 170692 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 18 |
Gene Aliases: |
ADAMTS21, KNO2, MMCAT |
Location: |
Chr.16:77282128-77435114 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADAMTS18 | NM_001326358.1 | NP_001313287.1 | ||
NM_199355.3 | NP_955387.1 | |||
XM_011522923.1 | XP_011521225.1 | |||
XM_011522924.1 | XP_011521226.1 | |||
XM_017022988.1 | XP_016878477.1 | |||
XM_017022989.1 | XP_016878478.1 | |||
AJ311903.1 | CAC83612.1 | |||
AK122677.1 | BAC85503.1 | |||
AK131403.1 | BAD18550.1 | |||
AY172951.1 | AAO45695.1 | |||
BC063283.1 | AAH63283.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056676 | Chr.16:77105785 - 77820951 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv1055234 | Chr.16:76969529 - 77433389 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv3027n100 | Chr.16:76614676 - 77688151 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1065714 | Chr.16:77251762 - 77351621 on Build GRCh38 | Loss |
![]() |
esv2761916 | Chr.16:77243108 - 77521936 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
