Assay Details
Target Gene Details
Entrez Gene ID: | 9683 |
Gene Name: | NEDD4 binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.16:48538726-48610214 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N4BP1 | NM_153029.3 | NP_694574.3 | ||
| XM_011523482.1 | XP_011521784.1 | |||
| AB014515.1 | ||||
| AK001464.1 | ||||
| AK026937.1 | ||||
| BC108288.1 | AAI08289.1 | |||
| BC152472.1 |
More Information
Set Membership: |
|

Genomic Map