Assay Details
Target Gene Details
Entrez Gene ID: | 55718 |
Gene Name: | RNA polymerase III subunit E |
Gene Aliases: |
RPC5, SIN |
Location: |
Chr.16:22297366-22335103 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3E | NM_001258033.1 | NP_001244962.1 | ||
| NM_001258034.1 | NP_001244963.1 | |||
| NM_001258035.1 | NP_001244964.1 | |||
| NM_001258036.1 | NP_001244965.1 | |||
| NM_018119.3 | NP_060589.1 | |||
| NR_047581.1 | ||||
| XM_006721063.2 | XP_006721126.1 | |||
| XM_011545892.1 | XP_011544194.1 | |||
| XM_011545893.2 | XP_011544195.1 | |||
| XM_017023443.1 | XP_016878932.1 | |||
| AB040885.1 | BAA95976.1 | |||
| AK001371.1 | BAA91655.1 | |||
| AK023160.1 | BAB14437.1 | |||
| AK023231.1 | BAB14481.1 | |||
| AK026645.1 | ||||
| AK130661.1 | ||||
| AK222671.1 | ||||
| AK222789.1 | BAD96509.1 | |||
| AK293300.1 | ||||
| AK296813.1 | ||||
| AK300735.1 | ||||
| AY092085.1 | AAM18215.1 | |||
| BC000285.1 | AAH00285.1 | |||
| DB047155.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638204 | Chr.16:22296113 - 22352456 on Build GRCh38 | Gain |
|
| nsv952951 | Chr.16:22299380 - 22329279 on Build GRCh38 | Deletion |
|
| esv2758637 | Chr.16:22282803 - 22760988 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map