Assay Details
Target Gene Details
Entrez Gene ID: | 84937 |
Gene Name: | zinc and ring finger 1, E3 ubiquitin protein ligase |
Gene Aliases: |
NIN283 |
Location: |
Chr.16:74999017-75110994 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNRF1 | NM_032268.4 | NP_115644.1 | ||
XM_011523392.1 | XP_011521694.1 | |||
XM_017023793.1 | XP_016879282.1 | |||
AF378524.1 | AAK69753.1 | |||
AK294434.1 | ||||
AL136903.1 | CAB66837.1 | |||
AL834440.1 | CAD39100.1 | |||
BC007235.1 | AAH07235.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833280 | Chr.16:74821913 - 75043329 on Build GRCh38 | Loss |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
