Assay Details
Target Gene Details
Entrez Gene ID: | 54739 |
Gene Name: | XIAP associated factor 1 |
Gene Aliases: |
BIRC4BP, HSXIAPAF1, XIAPAF1 |
Location: |
Chr.17:6755411-6775647 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XAF1 | NM_017523.3 | NP_059993.2 | ||
| NM_199139.2 | NP_954590.1 | |||
| NR_046396.1 | ||||
| NR_046397.1 | ||||
| NR_046398.1 | ||||
| XM_005256706.4 | XP_005256763.1 | |||
| XM_005256708.2 | XP_005256765.1 | |||
| XM_011523945.2 | XP_011522247.1 | |||
| XM_011523946.2 | XP_011522248.1 | |||
| XM_011523947.2 | XP_011522249.1 | |||
| XM_011523948.1 | XP_011522250.1 | |||
| XM_017024780.1 | XP_016880269.1 | |||
| AK056908.1 | ||||
| AK290276.1 | ||||
| AK292710.1 | ||||
| AK292848.1 | ||||
| BC032776.1 | AAH32776.1 | |||
| BC073156.1 | AAH73156.1 | |||
| BM475564.1 | ||||
| BX648588.1 | ||||
| BX649188.1 | CAE46206.1 | |||
| EF028165.1 | ||||
| EF028166.1 | ||||
| X99699.1 | CAA68030.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059072 | Chr.17:6727117 - 6876318 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map