Assay Details
Target Gene Details
Entrez Gene ID: | 162333 |
Gene Name: | membrane associated ring-CH-type finger 10 |
Gene Aliases: |
MARCH-X, RNF190 |
Location: |
Chr.17:62701314-62808381 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARCH10 | NM_001100875.2 | NP_001094345.1 | ||
| NM_001288779.1 | NP_001275708.1 | |||
| NM_001288780.1 | NP_001275709.1 | |||
| NM_152598.3 | NP_689811.2 | |||
| XM_005257095.2 | XP_005257152.2 | |||
| XM_005257096.2 | XP_005257153.2 | |||
| XM_005257097.3 | XP_005257154.2 | |||
| XM_005257098.2 | XP_005257155.2 | |||
| XM_005257100.2 | XP_005257157.2 | |||
| XM_005257102.2 | XP_005257159.2 | |||
| XM_005257103.1 | XP_005257160.1 | |||
| XM_005257106.4 | XP_005257163.2 | |||
| XM_011524435.1 | XP_011522737.1 | |||
| XM_011524436.1 | XP_011522738.1 | |||
| XM_011524437.1 | XP_011522739.1 | |||
| AK093076.1 | BAC04044.1 | |||
| AK122947.1 | ||||
| BC035021.1 | AAH35021.1 | |||
| BX105355.1 | ||||
| BX648705.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2115 | Chr.17:62692811 - 62726853 on Build GRCh38 | Insertion |
|
| nsv516921 | Chr.17:62663603 - 62769297 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map