Assay Details
Target Gene Details
Entrez Gene ID: | 54739 |
Gene Name: | XIAP associated factor 1 |
Gene Aliases: |
BIRC4BP, HSXIAPAF1, XIAPAF1 |
Location: |
Chr.17:6755411-6775647 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XAF1 | XM_005256706.4 | XP_005256763.1 | ||
| XM_005256708.2 | XP_005256765.1 | |||
| AK056908.1 | ||||
| BX649188.1 | CAE46206.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059072 | Chr.17:6727117 - 6876318 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79926030] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map