Assay Details
Target Gene Details
Entrez Gene ID: | 101927018 |
Gene Name: | uncharacterized LOC101927018 |
Gene Aliases: |
- |
Location: |
Chr.17:28926275-28944749 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927018 | NR_110803.1 |
Target Gene Details
Entrez Gene ID: | 57649 |
Gene Name: | PHD finger protein 12 |
Gene Aliases: |
PF1 |
Location: |
Chr.17:28905250-28951490 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHF12 | NM_001033561.1 | NP_001028733.1 | ||
| NM_001290131.1 | NP_001277060.1 | |||
| NM_020889.2 | NP_065940.1 | |||
| XM_005258015.3 | XP_005258072.1 | |||
| XM_011525079.2 | XP_011523381.1 | |||
| XM_011525082.2 | XP_011523384.1 | |||
| XM_011525083.2 | XP_011523385.1 | |||
| XM_017024903.1 | XP_016880392.1 | |||
| XM_017024904.1 | XP_016880393.1 | |||
| AK160370.1 | BAD18713.1 | |||
| AK291420.1 | ||||
| AK294056.1 | ||||
| AL161953.1 | ||||
| AY030283.1 | AAK38349.1 | |||
| BC110882.1 | AAI10883.1 | |||
| BC121043.1 | ||||
| BC121044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457719 | Chr.17:28867616 - 29014758 on Build GRCh38 | Loss |
|
| nsv833408 | Chr.17:28913470 - 29105450 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map