Assay Details
Target Gene Details
Entrez Gene ID: | 124817 |
Gene Name: | cyclin N-terminal domain containing 1 |
Gene Aliases: |
CNTD |
Location: |
Chr.17:42798667-42811587 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTD1 | NM_173478.2 | NP_775749.2 | ||
| XM_005257043.4 | XP_005257100.1 | |||
| XM_005257044.4 | XP_005257101.1 | |||
| XM_005257045.4 | XP_005257102.1 | |||
| XM_006721690.2 | XP_006721753.1 | |||
| XM_011524310.2 | XP_011522612.1 | |||
| XM_011524311.2 | XP_011522613.1 | |||
| XM_017024171.1 | XP_016879660.1 | |||
| AK097456.1 | BAC05060.1 | |||
| AK302094.1 | ||||
| BC026187.2 | AAH26187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833450 | Chr.17:42660985 - 42825144 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs575351771] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map