Assay Details
Target Gene Details
Entrez Gene ID: | 84073 |
Gene Name: | MYCBP associated protein |
Gene Aliases: |
AMAP-1, AMAP1 |
Location: |
Chr.17:50507778-50531502 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCBPAP | NM_032133.4 | NP_115509.4 | ||
| XM_005257725.3 | XP_005257782.1 | |||
| XM_005257726.2 | XP_005257783.1 | |||
| XM_011525342.1 | XP_011523644.1 | |||
| XM_011525343.1 | XP_011523645.1 | |||
| XM_017025205.1 | XP_016880694.1 | |||
| AB083068.1 | BAB88692.1 | |||
| AK303041.1 | ||||
| AK314052.1 | ||||
| AL832225.1 | CAH56203.1 | |||
| BC028393.2 | AAH28393.1 | |||
| BG718526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2082 | Chr.17:50501715 - 50528088 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map