Assay Details
Target Gene Details
Entrez Gene ID: | 9094 |
Gene Name: | unc-119 lipid binding chaperone |
Gene Aliases: |
HRG4, IMD13, POC7, POC7A |
Location: |
Chr.17:28546707-28552633 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC119 | NM_005148.3 | NP_005139.1 | ||
| NM_054035.2 | NP_473376.1 | |||
| XM_011525459.2 | XP_011523761.1 | |||
| XM_017025289.1 | XP_016880778.1 | |||
| AB593014.1 | ||||
| AF028788.1 | AAD01875.1 | |||
| AF028789.1 | AAD01876.1 | |||
| AK124531.1 | 1 | 1170 | ||
| AK292329.1 | ||||
| BC027176.1 | AAH27176.1 | |||
| BM848267.1 | ||||
| BX641031.1 | 1 | 1093 | CAE46019.1 | |
| U40998.1 | AAC50360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|
| nsv1064958 | Chr.17:28490519 - 28565886 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map