Assay Details
Target Gene Details
Entrez Gene ID: | 81558 |
Gene Name: | family with sequence similarity 117 member A |
Gene Aliases: |
- |
Location: |
Chr.17:49710325-49764156 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM117A | NM_030802.3 | NP_110429.1 | ||
| XM_017025182.1 | XP_016880671.1 | |||
| AF123073.1 | AAK01477.1 | |||
| AK302376.1 | ||||
| BC037572.1 | AAH37572.1 | |||
| BC065199.1 | AAH65199.1 |

Genomic Map