Assay Details
Target Gene Details
Entrez Gene ID: | 56919 |
Gene Name: | DEAH-box helicase 33 |
Gene Aliases: |
DDX33 |
Location: |
Chr.17:5440912-5469060 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX33 | NM_001199699.1 | NP_001186628.1 | ||
| NM_020162.3 | NP_064547.2 | |||
| AK295760.1 |
Target Gene Details
Entrez Gene ID: | 105371506 |
Gene Name: | uncharacterized LOC105371506 |
Gene Aliases: |
- |
Location: |
Chr.17:5469092-5470360 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105371506 | NR_135644.1 | |||
| AW103155.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74973471] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map