Assay Details
Target Gene Details
Entrez Gene ID: | 284021 |
Gene Name: | mast cell immunoglobulin like receptor 1 |
Gene Aliases: |
Allergin-1, C17orf60, MCA-32, MCA32 |
Location: |
Chr.17:64449022-64474009 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MILR1 | XM_017024486.1 | 10 | 1349 | XP_016879975.1 |
More Information
Set Membership: |
|

Genomic Map