Assay Details
Target Gene Details
Entrez Gene ID: | 100505591 |
Gene Name: | leucine rich repeat containing 3C |
Gene Aliases: |
- |
Location: |
Chr.17:39927722-39944999 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC3C | XM_011524158.2 | XP_011522460.1 | ||
| AK091224.1 | 1 | 1691 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573064 | Chr.17:39896348 - 40029806 on Build GRCh38 | Gain |
|

Genomic Map