Assay Details
Target Gene Details
Entrez Gene ID: | 3060 |
Gene Name: | hypocretin neuropeptide precursor |
Gene Aliases: |
NRCLP1, OX, PPOX |
Location: |
Chr.17:42184060-42185452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCRT | NM_001524.1 | NP_001515.1 | ||
| AF041240.1 | AAC39600.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056904 | Chr.17:42025823 - 42363204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map