Assay Details
Target Gene Details
Entrez Gene ID: | 5175 |
Gene Name: | platelet and endothelial cell adhesion molecule 1 |
Gene Aliases: |
CD31, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, endoCAM |
Location: |
Chr.17:64319415-64413844 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PECAM1 | NM_000442.4 | 16 | 4901 | NP_000433.4 |
| AK091419.1 | 1 | 735 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066968 | Chr.17:64215762 - 64332226 on Build GRCh38 | Gain |
|
| nsv833513 | Chr.17:64235366 - 64396432 on Build GRCh38 | Loss |
|
| nsv524852 | Chr.17:64215039 - 64329779 on Build GRCh38 | Gain |
|
| nsv833512 | Chr.17:64210750 - 64396432 on Build GRCh38 | Gain+Loss |
|
| nsv509666 | Chr.17:64254455 - 64396455 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map