Assay Details
Target Gene Details
Entrez Gene ID: | 124773 |
Gene Name: | chromosome 17 open reading frame 64 |
Gene Aliases: |
- |
Location: |
Chr.17:60421091-60431426 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C17orf64 | XM_005257033.4 | XP_005257090.1 | ||
| XM_005257034.4 | XP_005257091.1 |
Target Gene Details
Entrez Gene ID: | 84669 |
Gene Name: | ubiquitin specific peptidase 32 |
Gene Aliases: |
NY-REN-60, USP10 |
Location: |
Chr.17:60177231-60422470 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP32 | XM_011525371.1 | XP_011523673.1 | ||
| XM_011525372.1 | XP_011523674.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3273n100 | Chr.17:60293516 - 60519251 on Build GRCh38 | Gain |
|
| nsv519929 | Chr.17:60311418 - 60431257 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map