Assay Details
Target Gene Details
Entrez Gene ID: | 83990 |
Gene Name: | BRCA1 interacting protein C-terminal helicase 1 |
Gene Aliases: |
BACH1, FANCJ, OF |
Location: |
Chr.17:61679186-61864120 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRIP1 | NM_032043.2 | NP_114432.2 | ||
| XM_011525332.2 | XP_011523634.1 | |||
| XM_011525333.2 | XP_011523635.1 | |||
| XM_011525334.2 | XP_011523636.1 | |||
| XM_011525335.2 | XP_011523637.1 | |||
| XM_011525336.2 | XP_011523638.1 | |||
| XM_011525337.2 | XP_011523639.1 | |||
| XM_011525338.2 | XP_011523640.1 | |||
| XM_011525340.2 | XP_011523642.1 | |||
| XM_017025200.1 | XP_016880689.1 | |||
| XM_017025201.1 | XP_016880690.1 | |||
| XM_017025202.1 | XP_016880691.1 | |||
| XM_017025203.1 | XP_016880692.1 | |||
| AF360549.1 | AAK38111.1 | |||
| AK074713.1 | BAC11156.1 | |||
| BC101472.1 | AAI01473.1 | |||
| BC101474.1 | AAI01475.1 |
More Information
Set Membership: |
|

Genomic Map