Assay Details
Target Gene Details
Entrez Gene ID: | 15 |
Gene Name: | aralkylamine N-acetyltransferase |
Gene Aliases: |
DSPS, SNAT |
Location: |
Chr.17:76453351-76470797 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AANAT | NM_001166579.1 | NP_001160051.1 | ||
| XM_017024259.1 | 1 | 1184 | XP_016879748.1 | |
| BC092430.1 | AAH92430.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058263 | Chr.17:76466514 - 76530556 on Build GRCh38 | Gain |
|
| nsv833549 | Chr.17:76365444 - 76527890 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map