Assay Details
Target Gene Details
Entrez Gene ID: | 1655 |
Gene Name: | DEAD-box helicase 5 |
Gene Aliases: |
G17P1, HLR1, HUMP68, p68 |
Location: |
Chr.17:64498254-64507038 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX5 | NM_001320595.1 | 14 | 3122 | NP_001307524.1 |
| NM_001320596.1 | 14 | 3011 | NP_001307525.1 | |
| NM_001320597.1 | 13 | 2862 | NP_001307526.1 | |
| NM_004396.4 | 13 | 3097 | NP_004387.1 | |
| AL133585.1 | 1 | 244 | ||
| CN280686.1 | 1 | 407 |
More Information
Set Membership: |
|

Genomic Map