Assay Details
Target Gene Details
Entrez Gene ID: | 100506755 |
Gene Name: | mir-497-195 cluster host gene |
Gene Aliases: |
MIR195HG |
Location: |
Chr.17:7015818-7019654 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR497HG | NR_038310.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv952117 | Chr.17:7014482 - 7028381 on Build GRCh38 | Deletion |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
| nsv1121929 | Chr.17:7017681 - 7021081 on Build GRCh38 | Deletion |
|

Genomic Map