Assay Details
Target Gene Details
Entrez Gene ID: | 57551 |
Gene Name: | TAO kinase 1 |
Gene Aliases: |
KFC-B, MAP3K16, MARKK, PSK-2, PSK2, TAO1, hKFC-B, hTAOK1 |
Location: |
Chr.17:29390925-29551904 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TAOK1 | NM_020791.2 | NP_065842.1 | ||
NM_025142.1 | NP_079418.1 | |||
BC133039.1 | ||||
BC144067.1 | ||||
BE772351.1 | ||||
CA395289.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
