Assay Details
Target Gene Details
Entrez Gene ID: | 201181 |
Gene Name: | zinc finger protein 385C |
Gene Aliases: |
- |
Location: |
Chr.17:42025576-42065668 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF385C | XM_006721751.3 | XP_006721814.1 | ||
| XM_011524475.2 | 3 | 2019 | XP_011522777.1 | |
| XM_011524476.1 | 3 | 1970 | XP_011522778.1 | |
| XM_011524477.2 | 3 | 2377 | XP_011522779.1 | |
| XM_011524478.2 | 3 | 2642 | XP_011522780.1 | |
| XM_011524479.2 | XP_011522781.1 | |||
| XM_011524482.2 | XP_011522784.1 | |||
| XM_011524483.2 | XP_011522785.1 | |||
| XM_017024314.1 | XP_016879803.1 | |||
| XM_017024315.1 | XP_016879804.1 | |||
| XM_017024316.1 | XP_016879805.1 | |||
| XM_017024317.1 | XP_016879806.1 | |||
| XM_017024318.1 | XP_016879807.1 | |||
| XM_017024319.1 | XP_016879808.1 | |||
| XM_017024320.1 | XP_016879809.1 | |||
| XM_017024321.1 | XP_016879810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056904 | Chr.17:42025823 - 42363204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map