Assay Details
Target Gene Details
Entrez Gene ID: | 15 |
Gene Name: | aralkylamine N-acetyltransferase |
Gene Aliases: |
DSPS, SNAT |
Location: |
Chr.17:76453351-76470797 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AANAT | NM_001166579.1 | NP_001160051.1 | ||
| BC092430.1 | AAH92430.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833549 | Chr.17:76365444 - 76527890 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140643318] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map