Assay Details
Target Gene Details
Entrez Gene ID: | 25984 |
Gene Name: | keratin 23 |
Gene Aliases: |
CK23, HAIK1, K23 |
Location: |
Chr.17:40922696-40937643 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT23 | NM_001282433.1 | NP_001269362.1 | ||
| NM_015515.4 | NP_056330.3 | |||
| XM_005257200.4 | XP_005257257.1 | |||
| XM_011524595.1 | XP_011522897.1 | |||
| AF102848.1 | AAK00050.1 | |||
| AK002047.1 | BAA92054.1 | |||
| AK225866.1 | ||||
| AK289449.1 | ||||
| AK302109.1 | ||||
| AK310880.1 | ||||
| AK314604.1 | ||||
| AL117538.1 | CAB55984.2 | |||
| BC028356.2 | AAH28356.1 | |||
| DA846104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9550 | Chr.17:40888389 - 41135577 on Build GRCh38 | Loss |
|
| esv3640559 | Chr.17:40897964 - 41118942 on Build GRCh38 | Gain |
|
| nsv574999 | Chr.17:40910399 - 40979621 on Build GRCh38 | Loss |
|
| esv2761957 | Chr.17:40931731 - 41024070 on Build GRCh38 | Gain |
|
| esv3640560 | Chr.17:40902724 - 41042362 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs150557606] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map