Assay Details
Target Gene Details
Entrez Gene ID: | 101928514 |
Gene Name: | uncharacterized LOC101928514 |
Gene Aliases: |
- |
Location: |
Chr.17:76799044-76807102 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928514 | NR_110837.1 | 2 | 1047 | |
| AK055500.1 | 2 | 1047 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833550 | Chr.17:76712038 - 76886290 on Build GRCh38 | Loss |
|
| nsv1059991 | Chr.17:76802292 - 76896772 on Build GRCh38 | Loss |
|

Genomic Map