Assay Details
Target Gene Details
Entrez Gene ID: | 84073 |
Gene Name: | MYCBP associated protein |
Gene Aliases: |
AMAP-1, AMAP1 |
Location: |
Chr.17:50507778-50531502 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYCBPAP |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2082 | Chr.17:50501715 - 50528088 on Build GRCh38 | Insertion |
|

Genomic Map