Assay Details
Target Gene Details
Entrez Gene ID: | 8894 |
Gene Name: | eukaryotic translation initiation factor 2 subunit beta |
Gene Aliases: |
EIF2, EIF2B, EIF2beta, PPP1R67, eIF-2-beta |
Location: |
Chr.20:34088298-34112356 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2S2 | NM_001316363.1 | NP_001303292.1 | ||
| NM_001316364.1 | NP_001303293.1 | |||
| NM_003908.4 | NP_003899.2 | |||
| XM_017028118.1 | XP_016883607.1 | |||
| AB451237.1 | ||||
| AB451362.1 | ||||
| AK313025.1 | ||||
| BC000461.2 | AAH00461.1 | |||
| BC000934.2 | AAH00934.1 | |||
| BC007888.2 | AAH07888.1 | |||
| BT009795.1 | AAP88797.1 | |||
| BX418731.2 | ||||
| BX429051.2 | ||||
| CR456734.1 | CAG33015.1 | |||
| M29536.1 | AAA52383.1 |
More Information
Set Membership: |
|

Genomic Map