Assay Details
Target Gene Details
Entrez Gene ID: | 84677 |
Gene Name: | Down syndrome critical region 8 |
Gene Aliases: |
C21orf65, CT25.1a, CT25.1b, MMA-1, MMA-1a, MMA-1b, MMA1, MTAG2 |
Location: |
Chr.21:38121451-38156511 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSCR8 | NR_026838.1 | |||
| NR_026839.1 | ||||
| NR_026840.1 | ||||
| NR_026841.1 | ||||
| NR_026842.1 | ||||
| AA707621.1 | ||||
| AA770014.1 | ||||
| AF321193.1 | AAK52673.1 | |||
| AF426256.1 | AAM53512.1 | |||
| AJ001904.1 | ||||
| AJ297914.2 | CAC82786.1 | |||
| AJ301615.2 | CAD29791.1 | |||
| AJ305085.2 | CAH03994.1 | |||
| AJ306839.2 | CAC84766.1 | |||
| AJ306840.2 | CAC84765.1 | |||
| AJ783421.1 | CAH03845.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834097 | Chr.21:38111167 - 38205081 on Build GRCh38 | Gain |
|
| nsv1058886 | Chr.21:37969257 - 38427716 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map